Merck
  • Exploring olfactory receptor family 7 subfamily C member 1 as a novel oral cancer stem cell target for immunotherapy.
Exploring olfactory receptor family 7 subfamily C member 1 as a novel oral cancer stem cell target for immunotherapy.
Cancer science (2023-06-22)
Sho Miyamoto, Yoshihiko Hirohashi, Rena Morita, Akihiro Miyazaki, Kazuhiro Ogi, Takayuki Kanaseki, Kentaro Ide, Jumpei Shirakawa, Tomohide Tsukahara, Aiko Murai, Takashi Sasaya, Kazushige Koike, Shinichiro Kina, Toshihiro Kawano, Takahiro Goto, Edward Hosea Ntege, Yusuke Shimizu, Toshihiko Torigoe
ZUSAMMENFASSUNG
The mortality rate of oral cancer has not improved over the past three decades despite remarkable advances in cancer therapies. Oral cancers contain a subpopulation of cancer stem cells (CSCs) that share characteristics associated with normal stem cells, including self-renewal and multi-differentiation potential. CSCs are tumorigenic, play a critical role in cancer infiltration, recurrence, and distant metastasis, and significantly contribute to drug resistance to current therapeutic strategies, including immunotherapy. Cytotoxic CD8+ T lymphocytes (CTLs) are key immune cells that effectively recognize peptide antigens presented by the major histocompatibility complex class I molecules. Increasing evidence suggests that cancer antigen-specific targeting by CTLs effectively regulates CSCs that drive cancer progression. In this study, we utilized data from public domains and performed various bioassays on human oral squamous cell carcinoma clinical samples and cell lines, including HSC-2 and HSC-3, to investigate the potential role of olfactory receptor family 7 subfamily C member 1 (OR7C1), a seven transmembrane G-protein-coupled olfactory receptor that is also expressed in nonolfactory tissues and was previously reported as a novel marker and target of colon cancer initiating cell-targeted immunotherapy, in CSC-targeted treatment against oral cancer. We found that the OR7C1 gene was expressed only in oral CSCs, and that CTLs reacted with human leukocyte antigen-A24-restricted OR7C1 oral CSC-specific peptides. Taken together, our findings suggest that OR7C1 represents a novel target for potent CSC-targeted immunotherapy in oral cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-OR7C1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution