Merck
  • Characterization of calcium- and integrin-binding protein 1 (CIB1) knockout platelets: potential compensation by CIB family members.
Characterization of calcium- and integrin-binding protein 1 (CIB1) knockout platelets: potential compensation by CIB family members.
Thrombosis and haemostasis (2008-11-08)
Jan C Denofrio, Weiping Yuan, Brenda R Temple, Holly R Gentry, Leslie V Parise
ZUSAMMENFASSUNG
Platelet aggregation requires activation of the alphaIIbbeta3 integrin, an event regulated by the integrin cytoplasmic tails. CIB1 binds to the cytoplasmic tail of the integrin alphaIIb subunit. Previous over-expression and knockdown studies in murine megakaryocytes demonstrated that CIB1 inhibits integrin alphaIIbbeta3 activation. Here we analyzed Cib1(-/-) mice to determine the function of CIB1 in platelets in vitro and in vivo. We found that although these mice had no overt platelet phenotype, mRNA level of CIB1 homolog CIB3 was increased in Cib1(-/-) megakaryocytes. In vitro binding experiments showed that recombinant CIB1, -2 and -3 bound specifically to an alphaIIb cytoplasmic tail peptide. Subsequent protein modeling experiments indicated that CIBs 1-3 each have a highly conserved hydrophobic binding pocket. Therefore, the potential exists for compensation for the loss of CIB1 by these CIB family members, thereby preventing pathologic thrombus formation in Cib1(-/-) mice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Harnstoff, meets USP testing specifications
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%