Merck
  • Oxidative stress and regulation of adipogenic differentiation capacity by sirtuins in adipose stem cells derived from female patients of advancing age.
Oxidative stress and regulation of adipogenic differentiation capacity by sirtuins in adipose stem cells derived from female patients of advancing age.
Scientific reports (2024-08-28)
Anne Bernhardt, Alan Jamil, Md Tanvir Morshed, Pia Ponnath, Veronika Gille, Nadine Stephan, Heinrich Sauer, Maria Wartenberg
ZUSAMMENFASSUNG
Patient age is critical for mesenchymal stem cell quality and differentiation capacity. We demonstrate that proliferation and adipogenic capacity of subcutaneous adipose stem cells (ASCs) from female patients declined with advanced age, associated with reduction in cell nucleus size, increase in nuclear lamina protein lamin B1/B2, and lamin A, upregulation of senescence marker p16INK4a and senescence-associated β-galactosidase activity. Adipogenic induction resulted in differentiation of adipocytes and upregulation of adipogenic genes CCAAT enhancer binding protein alpha, fatty acid binding protein 4, lipoprotein lipase, and peroxisome proliferator-activated receptor-γ, which was not affected by the Sirt-1 activator YK-3-237 or the Sirt-1 inhibitor EX-527. Protein expression of the stem cell markers Oct4 and Sox2 was not significantly downregulated with advanced patient age. Mitochondrial reactive oxygen species were increased in ASCs from old-aged patients, whereas protein expression of NADPH oxidases NOX1 and NOX4 was downregulated, and dual oxidase isoforms remained unchanged. Generation of nitric oxide and iNOS expression was downregulated. Protein expression of Sirt-1 and Sirt-3 decreased with patient age, whereas Sirt-2 and Sirt-5 remained unchanged. Induction of adipogenesis stimulated protein expression of Sirt-1 and Sirt-3, which was not affected upon pre-incubation with the Sirt-1-activator YK-3-237 or the Sirt-1-inhibitor EX-527. The Sirt-1 inhibitor Sirtinol downregulated adiponectin protein expression and the number of adipocytes, whereas YK-3-237 exerted stimulatory effects. In summary, our data demonstrate increased oxidative stress in ASCs of aging patients, and decline of adipogenic capacity due to Sirt-1- mediated adiponectin downregulation in elderly patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-Vinculin-Antikörper in Maus hergestellte Antikörper, clone hVIN-1, ascites fluid