Merck
  • Multifunctional magnetic nanocarriers for image-tagged SiRNA delivery to intact pancreatic islets.
Multifunctional magnetic nanocarriers for image-tagged SiRNA delivery to intact pancreatic islets.
Transplantation (2008-11-14)
Zdravka Medarova, Mohanraja Kumar, Shu-Wing Ng, Junzheng Yang, Natasha Barteneva, Natalia V Evgenov, Victoria Petkova, Anna Moore
ZUSAMMENFASSUNG
With the ultimate hope of finding a cure for diabetes, researches are looking into altering the genetic profile of the beta cell as a way to manage metabolic dysregulation. One of the most powerful new approaches for the directed regulation of gene expression uses the phenomenon of RNA interference. Here, we establish the feasibility of a novel technology centered around multifunctional magnetic nanocarriers, which concurrently deliver siRNA to intact pancreatic islets and can be detected by magnetic resonance and optical imaging. In the proof-of-principle studies described here, we demonstrate that, after in vitro incubation, magnetic nanoparticles carrying siRNA designed to target the model gene for enhanced green fluorescent protein are efficiently taken up by murine pancreatic islets, derived from egfp transgenic animals. This uptake can be visualized by magnetic resonance imaging and near-infrared fluorescence optical imaging and results in suppression of the target gene. These results illustrate the value of our approach in overcoming the challenges associated with genetic modification of intact pancreatic islets in a clinically acceptable manner. Furthermore, an added advantage of our technology derives from the combined capability of our magnetic nanoparticles for siRNA delivery and magnetic labeling of pancreatic islets.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tris(2-carboxyethyl)phosphin -hydrochlorid, powder
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Tris(2-carboxyethyl)phosphin -hydrochlorid, BioUltra, ≥98% (NMR)
Sigma-Aldrich
L-Glutathion, oxidiert, ≥98% (HPLC)
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathion, oxidiert, ≥98%, lyophilized powder
Sigma-Aldrich
L-Glutathion, oxidiert, BioXtra, ≥98%