Merck
Elevated GRHL2 Imparts Plasticity in ER-Positive Breast Cancer Cells.
Cancers (2024-08-31)
Christy Zheng, Kaelyn O Allen, Tianrui Liu, Natalia M Solodin, Mark B Meyer, Kelley Salem, Phillipos K Tsourkas, Sean J McIlwain, Jessica M Vera, Erika R Cromwell, Mary Szatkowski Ozers, Amy M Fowler, Elaine T Alarid
ZUSAMMENFASSUNG
Estrogen receptor (ER)-positive breast cancer is characterized by late recurrences following initial treatment. The epithelial cell fate transcription factor Grainyhead-like protein 2 (GRHL2) is overexpressed in ER-positive breast cancers and is linked to poorer prognosis as compared to ER-negative breast cancers. To understand how GRHL2 contributes to progression, GRHL2 was overexpressed in ER-positive cells. We demonstrated that elevated GRHL2 imparts plasticity with stem cell- and dormancy-associated traits. RNA sequencing and immunocytochemistry revealed that high GRHL2 not only strengthens the epithelial identity but supports a hybrid epithelial to mesenchymal transition (EMT). Proliferation and tumor studies exhibited a decrease in growth and an upregulation of dormancy markers, such as NR2F1 and CDKN1B. Mammosphere assays and flow cytometry revealed enrichment of stem cell markers CD44 and ALDH1, and increased self-renewal capacity. Cistrome analyses revealed a change in transcription factor motifs near GRHL2 sites from developmental factors to those associated with disease progression. Together, these data support the idea that the plasticity and properties induced by elevated GRHL2 may provide a selective advantage to explain the association between GRHL2 and breast cancer progression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Puromycin -dihydrochlorid aus Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
Trypsin-Inhibitor aus Glycine max (soybean), solution, sterile-filtered, suitable for, suitable for cell culture
Sigma-Aldrich
Anti-GRHL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution