Merck
  • Conjunctival interleukin-13 expression in mucous membrane pemphigoid and functional effects of interleukin-13 on conjunctival fibroblasts in vitro.
Conjunctival interleukin-13 expression in mucous membrane pemphigoid and functional effects of interleukin-13 on conjunctival fibroblasts in vitro.
The American journal of pathology (2009-11-17)
Valerie P J Saw, Ifeoma Offiah, Robin J Dart, Grazyna Galatowicz, John K G Dart, Julie T Daniels, Virginia L Calder
ZUSAMMENFASSUNG
Interleukin-13 (IL-13) is the dominant effector cytokine of fibrosis in pulmonary and liver disease. Excessive conjunctival fibrosis in the immunobullous disease ocular mucous membrane pemphigoid (MMP) causes blindness; the pathogenesis of scarring in this disease is incompletely understood. To determine whether IL-13 is involved in conjunctival fibrosis in MMP, we studied the expression of IL-13 in ocular MMP patients before and after systemic immunosuppression and examined the effects of IL-13 on normal human conjunctival fibroblasts. We found high stromal cell expression of IL-13 in active ocular MMP by immunohistochemistry; 80% of these cells were CD3-positive T cells. Following immunosuppression, in clinically uninflamed, treated, ocular MMP patients, the number of IL-13 positive cells was significantly reduced, but this was still fourfold greater than in normal conjunctiva. IL-13 stimulated collagen lattice contraction and migration, and decreased production of mmp-3 and mmp-10 by human conjunctival fibroblasts. The addition of T cell culture supernatant to IL-13 synergistically augmented fibroblast migration. IL-13 also up-regulated surface expression of HLA-DR, CD80, CD40, and CD154 by conjunctival fibroblasts, suggesting a potential mechanism for fibroblast-T cell cross talk, via which fibroblasts may actively engage in perpetuating chronic inflammation and continued fibrosis. Together, these findings suggest that IL-13 is involved in conjunctival fibrosis in MMP, and that IL-13 has both profibrotic and pro-inflammatory effects on human conjunctival fibroblasts.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
Albumin aus Rinderserum, powder, BioXtra
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, suitable for (for molecular biology), Non-acetylated, Molecular Biology
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Albumin aus Rinderserum, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, pH <5.0, powder
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, low endotoxin, pH 7, ≥98%