Merck
Antiepileptogenic effects of trilostane in the kainic acid model of temporal lobe epilepsy.
Epilepsia (2023-03-03)
Anna Maria Costa, Mohammad Gol, Chiara Lucchi, Giuseppe Biagini
ZUSAMMENFASSUNG
Epileptogenesis after status epilepticus (SE) has a faster onset in rats treated to reduce brain levels of the anticonvulsant neurosteroid allopregnanolone with the 5α-reductase inhibitor finasteride; however, it still has to be evaluated whether treatments aimed at increasing allopregnanolone levels could result in the opposite effect of delaying epileptogenesis. This possibility could be tested using the peripherally active inhibitor of 3β-hydroxysteroid dehydrogenase/Δ5-4 isomerase trilostane, which has been shown repeatedly to increase allopregnanolone levels in the brain. Trilostane (50 mg/kg) was administered subcutaneously once daily for up to six consecutive days, starting 10 min after intraperitoneal administration of kainic acid (15 mg/kg). Seizures were evaluated by video-electrocorticographic recordings for 70 days maximum, and endogenous neurosteroid levels were assessed by liquid chromatography-electrospray tandem mass spectrometry. Immunohistochemical staining was performed to evaluate the presence of brain lesions. Trilostane did not alter the latency of kainic acid-induced SE onset or its overall duration. When compared to the vehicle-treated group, rats receiving six daily trilostane injections presented a remarkable delay of the first spontaneous electrocorticographic seizure and subsequent tonic-clonic spontaneous recurrent seizures (SRSs). Conversely, rats treated with only the first trilostane injection during SE did not differ from vehicle-treated rats in developing the SRSs. Notably, trilostane did not modify neuronal cell densities or the overall damage in the hippocampus. In comparison to the vehicle group, repeated administration of trilostane significantly decreased the activated microglia morphology in the subiculum. As expected, allopregnanolone and other neurosteroid levels were remarkably increased in the hippocampus and neocortex of rats treated for 6 days with trilostane, but pregnanolone was barely detectable. Neurosteroids returned to basal levels after a week of trilostane washout. Overall, these results suggest that trilostane led to a remarkable increase in allopregnanolone brain levels, which was associated with protracted effects on epileptogenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-NeuN-Antikörper, Klon A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Monoklonaler Anti-GFAP-Antikörper (Glial Fibrillary Acidic Protein, Saures Gliafaserprotein) in Maus hergestellte Antikörper, clone G-A-5, ascites fluid