Merck
  • Split-CreERT2: temporal control of DNA recombination mediated by split-Cre protein fragment complementation.
Split-CreERT2: temporal control of DNA recombination mediated by split-Cre protein fragment complementation.
PloS one (2009-12-18)
Johannes Hirrlinger, Robert P Requardt, Ulrike Winkler, Franziska Wilhelm, Christine Schulze, Petra G Hirrlinger
ZUSAMMENFASSUNG
DNA recombination technologies such as the Cre/LoxP system advance modern biological research by allowing conditional gene regulation in vivo. However, the precise targeting of a particular cell type at a given time point has remained challenging since spatial specificity has so far depended exclusively on the promoter driving Cre recombinase expression. We have recently established split-Cre that allows DNA recombination to be controlled by coincidental activity of two promoters, thereby increasing spatial specificity of Cre-mediated DNA recombination. To allow temporal control of split-Cre-mediated DNA recombination we have now extended split-Cre by fusing split-Cre proteins with the tamoxifen inducible ERT2 domain derived from CreERT2. In the split-CreERT2 system, Cre-mediated DNA recombination is controlled by two expression cassettes as well as the time of tamoxifen application. By using two independent Cre-dependent reporters in cultured cells, the combination of NCre-ERT2+ERT2-CCre was identified as having the most favorable properties of all constructs tested, showing an induction ratio of about 10 and EC(50)-values for 4-hydroxy-tamoxifen of 10 nM to 70 nM. These characteristics of split-CreERT2 in vitro indicate that split-CreERT2 will be well suited for inducing DNA recombination in living mice harboring LoxP-flanked alleles. In this way, split-CreERT2 will provide a new tool of modern genetics allowing spatial and temporal precise genetic access to cell populations defined by the simultaneous activity of two promoters.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Natriumpyruvat, BioXtra, ≥99%