- txci-ATAC-seq: a massive-scale single-cell technique to profile chromatin accessibility.
txci-ATAC-seq: a massive-scale single-cell technique to profile chromatin accessibility.
Genome biology (2024-03-23)
Hao Zhang, Ryan M Mulqueen, Natalie Iannuzo, Dominique O Farrera, Francesca Polverino, James J Galligan, Julie G Ledford, Andrew C Adey, Darren A Cusanovich
PMID38519979
ZUSAMMENFASSUNG
We develop a large-scale single-cell ATAC-seq method by combining Tn5-based pre-indexing with 10× Genomics barcoding, enabling the indexing of up to 200,000 nuclei across multiple samples in a single reaction. We profile 449,953 nuclei across diverse tissues, including the human cortex, mouse brain, human lung, mouse lung, mouse liver, and lung tissue from a club cell secretory protein knockout (CC16-/-) model. Our study of CC16-/- nuclei uncovers previously underappreciated technical artifacts derived from remnant 129 mouse strain genetic material, which cause profound cell-type-specific changes in regulatory elements near many genes, thereby confounding the interpretation of this commonly referenced mouse model.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution