Merck
  • Terminal differentiation of villus tip enterocytes is governed by distinct Tgfβ superfamily members.
Terminal differentiation of villus tip enterocytes is governed by distinct Tgfβ superfamily members.
EMBO reports (2023-07-26)
Linda Berková, Hassan Fazilaty, Qiutan Yang, Jan Kubovčiak, Monika Stastna, Dusan Hrckulak, Martina Vojtechova, Tosca Dalessi, Michael David Brügger, George Hausmann, Prisca Liberali, Vladimir Korinek, Konrad Basler, Tomas Valenta
ZUSAMMENFASSUNG
The protective and absorptive functions of the intestinal epithelium rely on differentiated enterocytes in the villi. The differentiation of enterocytes is orchestrated by sub-epithelial mesenchymal cells producing distinct ligands along the villus axis, in particular Bmps and Tgfβ. Here, we show that individual Bmp ligands and Tgfβ drive distinct enterocytic programs specific to villus zonation. Bmp4 is expressed from the centre to the upper part of the villus and activates preferentially genes connected to lipid uptake and metabolism. In contrast, Bmp2 is produced by villus tip mesenchymal cells and it influences the adhesive properties of villus tip epithelial cells and the expression of immunomodulators. Additionally, Tgfβ induces epithelial gene expression programs similar to those triggered by Bmp2. Bmp2-driven villus tip program is activated by a canonical Bmp receptor type I/Smad-dependent mechanism. Finally, we establish an organoid cultivation system that enriches villus tip enterocytes and thereby better mimics the cellular composition of the intestinal epithelium. Our data suggest that not only a Bmp gradient but also the activity of individual Bmp drives specific enterocytic programs.