Merck
HDAC1/2 inhibitor therapy improves multiple organ systems in aged mice.
iScience (2024-01-25)
Alessandra Tammaro, Eileen G Daniels, Iman M Hu, Kelly C 't Hart, Kim Reid, Rio P Juni, Loes M Butter, Goutham Vasam, Rashmi Kamble, Aldo Jongejan, Richard I Aviv, Joris J T H Roelofs, Eleonora Aronica, Reinier A Boon, Keir J Menzies, Riekelt H Houtkooper, Georges E Janssens
ZUSAMMENFASSUNG
Aging increases the risk of age-related diseases, imposing substantial healthcare and personal costs. Targeting fundamental aging mechanisms pharmacologically can promote healthy aging and reduce this disease susceptibility. In this work, we employed transcriptome-based drug screening to identify compounds emulating transcriptional signatures of long-lived genetic interventions. We discovered compound 60 (Cmpd60), a selective histone deacetylase 1 and 2 (HDAC1/2) inhibitor, mimicking diverse longevity interventions. In extensive molecular, phenotypic, and bioinformatic assessments using various cell and aged mouse models, we found Cmpd60 treatment to improve age-related phenotypes in multiple organs. Cmpd60 reduces renal epithelial-mesenchymal transition and fibrosis in kidney, diminishes dementia-related gene expression in brain, and enhances cardiac contractility and relaxation for the heart. In sum, our two-week HDAC1/2 inhibitor treatment in aged mice establishes a multi-tissue, healthy aging intervention in mammals, holding promise for therapeutic translation to promote healthy aging in humans.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-beta-Aktin-Antikörper, Klon 4C2, clone 4C2, from mouse