Merck
  • Excess pancreatic elastase alters acinar-β cell communication by impairing the mechano-signaling and the PAR2 pathways.
Excess pancreatic elastase alters acinar-β cell communication by impairing the mechano-signaling and the PAR2 pathways.
Cell metabolism (2023-06-21)
Giorgio Basile, Amedeo Vetere, Jiang Hu, Oluwaseun Ijaduola, Yi Zhang, Ka-Cheuk Liu, Amira M Eltony, Dario F De Jesus, Kazuki Fukuda, Grace Doherty, Colin A Leech, Oleg G Chepurny, George G Holz, Seok-Hyun Yun, Olov Andersson, Amit Choudhary, Bridget K Wagner, Rohit N Kulkarni
ZUSAMMENFASSUNG
Type 1 (T1D) or type 2 diabetes (T2D) are caused by a deficit of functional insulin-producing β cells. Thus, the identification of β cell trophic agents could allow the development of therapeutic strategies to counteract diabetes. The discovery of SerpinB1, an elastase inhibitor that promotes human β cell growth, prompted us to hypothesize that pancreatic elastase (PE) regulates β cell viability. Here, we report that PE is up-regulated in acinar cells and in islets from T2D patients, and negatively impacts β cell viability. Using high-throughput screening assays, we identified telaprevir as a potent PE inhibitor that can increase human and rodent β cell viability in vitro and in vivo and improve glucose tolerance in insulin-resistant mice. Phospho-antibody microarrays and single-cell RNA sequencing analysis identified PAR2 and mechano-signaling pathways as potential mediators of PE. Taken together, our work highlights PE as a potential regulator of acinar-β cell crosstalk that acts to limit β cell viability, leading to T2D.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Sigma-Aldrich
Anti-Phospho-Histon H3 (Ser10)-Antikörper, Mitosemarker, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-ELA3B antibody produced in mouse, clone 3H3, purified immunoglobulin, buffered aqueous solution