Merck
  • Role of Noradrenaline and Adrenoreceptors in Regulating Prostaglandin E2 Synthesis Cascade in Inflamed Endometrium of Pigs.
Role of Noradrenaline and Adrenoreceptors in Regulating Prostaglandin E2 Synthesis Cascade in Inflamed Endometrium of Pigs.
International journal of molecular sciences (2023-03-30)
Barbara Jana, Jarosław Całka, Michał Bulc, Krzysztof Witek
ZUSAMMENFASSUNG
In the inflamed uterus, the production and secretion of prostaglandins (PGs) and noradrenergic innervation pattern are changed. Receptor-based control of prostaglandin E2 (PGE2) production and secretion by noradrenaline during uterine inflammation is unknown. The aim of this study was to determine the role of α1-, α2- and β-adrenoreceptors (ARs) in noradrenaline-influenced PG-endoperoxidase synthase-2 (PTGS-2) and microsomal PTGE synthase-1 (mPTGES-1) protein levels in the inflamed pig endometrium, and in the secretion of PGE2 from this tissue. E. coli suspension (E. coli group) or saline (CON group) was injected into the uterine horns. Eight days later, severe acute endometritis developed in the E. coli group. Endometrial explants were incubated with noradrenaline and/or α1-, α2- and β-AR antagonists. In the CON group, noradrenaline did not significantly change PTGS-2 and mPTGES-1 protein expression and increased PGE2 secretion compared to the control values (untreated tissue). In the E. coli group, both enzyme expression and PGE2 release were stimulated by noradrenaline, and these values were higher versus the CON group. The antagonists of α1- and α2-AR isoforms and β-AR subtypes do not significantly alter the noradrenaline effect on PTGS-2 and mPTGES-1 protein levels in the CON group, compared to noradrenaline action alone. In this group, α1A-, α2B- and β2-AR antagonists partly eliminated noradrenaline-stimulated PGE2 release. Compared to the noradrenaline effect alone, α1A-, α1B-, α2A-, α2B-, β1-, β2- and β3-AR antagonists together with noradrenaline reduced PTGS-2 protein expression in the E. coli group. Such effects were also exerted in this group by α1A-, α1D-, α2A-, β2- and β3-AR antagonists with noradrenaline on mPTGES-1 protein levels. In the E. coli group, the antagonists of all isoforms of α1-ARs and subtypes of β-ARs as well as α2A-ARs together with noradrenaline decreased PGE2 secretion versus noradrenaline action alone. Summarizing, in the inflamed pig endometrium, α1(A, B)-, α2(A, B)- and β(1, 2, 3)-ARs mediate the noradrenaline stimulatory effect on PTGE-2 protein expression, while noradrenaline via α1(A, D)-, α2A- and β(2, 3)-ARs increases mPTGES-1 protein expression and α1(A, B, D)-, α2A- and β(1, 2, 3)-ARs are involved in PGE2 release. Data suggest that noradrenaline may indirectly affect the processes regulated by PGE2 by influencing its production. Pharmacological modulation of particular AR isoforms/subtypes can be used to change PGE2 synthesis/secretion to alleviate inflammation and improve uterine function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-GAPDH in Kaninchen hergestellte Antikörper, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution