Merck
Arginine reprograms metabolism in liver cancer via RBM39.
Cell (2023-10-08)
Dirk Mossmann, Christoph Müller, Sujin Park, Brendan Ryback, Marco Colombi, Nathalie Ritter, Diana Weißenberger, Eva Dazert, Mairene Coto-Llerena, Sandro Nuciforo, Lauriane Blukacz, Caner Ercan, Veronica Jimenez, Salvatore Piscuoglio, Fatima Bosch, Luigi M Terracciano, Uwe Sauer, Markus H Heim, Michael N Hall
ZUSAMMENFASSUNG
Metabolic reprogramming is a hallmark of cancer. However, mechanisms underlying metabolic reprogramming and how altered metabolism in turn enhances tumorigenicity are poorly understood. Here, we report that arginine levels are elevated in murine and patient hepatocellular carcinoma (HCC), despite reduced expression of arginine synthesis genes. Tumor cells accumulate high levels of arginine due to increased uptake and reduced arginine-to-polyamine conversion. Importantly, the high levels of arginine promote tumor formation via further metabolic reprogramming, including changes in glucose, amino acid, nucleotide, and fatty acid metabolism. Mechanistically, arginine binds RNA-binding motif protein 39 (RBM39) to control expression of metabolic genes. RBM39-mediated upregulation of asparagine synthesis leads to enhanced arginine uptake, creating a positive feedback loop to sustain high arginine levels and oncogenic metabolism. Thus, arginine is a second messenger-like molecule that reprograms metabolism to promote tumor growth.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
N-Acetyl-L-Cystein, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Aktin-Antikörper, Klon C4, ascites fluid, clone C4, Chemicon®