Merck
  • Distinct distribution of subplate neuron subtypes between the sensory cortices during the early postnatal period.
Distinct distribution of subplate neuron subtypes between the sensory cortices during the early postnatal period.
The Journal of comparative neurology (2024-02-26)
Minzi Chang, Sophia Nehs, Zheng Xu, Patrick O Kanold
ZUSAMMENFASSUNG
Subplate neurons (SpNs) are a heterogeneous neuronal population actively involved in early cortical circuit formation. In rodents, many SpNs survive and form layer 6b. The molecular heterogeneity of SpNs raises the question of whether different subpopulations of SpNs survive through the early postnatal period similarly and whether such diverse SpN populations in the auditory cortex (ACtx) share a common distribution pattern with other sensory systems. To address that, we investigated the expression pattern of multiple specific SpN markers in the ACtx, as well as in the visual (VCtx) and somatosensory (SCtx) cortices as controls, using complexin 3 (Cplx3) antibodies and different SpN-specific Cre-driver mice, such as connective tissue growth factor (CTGF), dopamine receptor D1 (Drd1a), and neurexophilin 4 (Nxph4). We focused on two early time windows in auditory development: (1) during the second postnatal week (PNW) before ear-canal opening and (2) during the third PNW after ear-canal opening. We compared the expression pattern of different SpN markers in ACtx with VCtx and SCtx. At both examined timepoints, Cplx3 and Nxph4 expressing SpNs form the largest and smallest population in the ACtx, respectively. Similar distribution patterns are observable in the VCtx and SCtx during the second PNW but not during the third PNW, for a higher proportion of Drd1a expressing SpNs is detected in the VCtx and CTGF expressing SpNs in the SCtx. This study suggests that different populations of SpNs might contribute differently to the development of individual sensory circuits.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen, ≥99%