Merck
Allergen exposure functionally alters influenza-specific CD4+ Th1 memory cells in the lung.
The Journal of experimental medicine (2023-09-12)
Mikel J Rüterbusch, Brian D Hondowicz, Kennidy K Takehara, Kurt B Pruner, Thomas S Griffith, Marion Pepper
ZUSAMMENFASSUNG
CD4+ lung-resident memory T cells (TRM) generated in response to influenza infection confer effective protection against subsequent viral exposures. Whether these cells can be altered by environmental antigens and cytokines released during heterologous, antigen-independent immune responses is currently unclear. We therefore investigated how influenza-specific CD4+ Th1 TRM in the lung are impacted by a subsequent Th2-inducing respiratory house dust mite (HDM) exposure. Although naïve influenza-specific CD4+ T cells in the lymph nodes do not respond to HDM, influenza-specific CD4+ TRM in the lungs do respond to a subsequent allergen exposure by decreasing expression of the transcription factor T-bet. This functional alteration is associated with decreased IFN-γ production upon restimulation and improved disease outcomes following heterosubtypic influenza challenge. Further investigation revealed that ST2 signaling in CD4+ T cells during allergic challenge is necessary to induce these changes in lung-resident influenza-specific CD4+ TRM. Thus, heterologous antigen exposure or ST2-signaling can drive persistent changes in CD4+ Th1 TRM populations and impact protection upon reinfection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen, ≥99%