Merck
  • Ketogenic diet promotes tumor ferroptosis but induces relative corticosterone deficiency that accelerates cachexia.
Ketogenic diet promotes tumor ferroptosis but induces relative corticosterone deficiency that accelerates cachexia.
Cell metabolism (2023-06-14)
Miriam Ferrer, Nicholas Mourikis, Emma E Davidson, Sam O Kleeman, Marta Zaccaria, Jill Habel, Rachel Rubino, Qing Gao, Thomas R Flint, Lisa Young, Claire M Connell, Michael J Lukey, Marcus D Goncalves, Eileen P White, Ashok R Venkitaraman, Tobias Janowitz
ZUSAMMENFASSUNG
Glucose dependency of cancer cells can be targeted with a high-fat, low-carbohydrate ketogenic diet (KD). However, in IL-6-producing cancers, suppression of the hepatic ketogenic potential hinders the utilization of KD as energy for the organism. In IL-6-associated murine models of cancer cachexia, we describe delayed tumor growth but accelerated cachexia onset and shortened survival in mice fed KD. Mechanistically, this uncoupling is a consequence of the biochemical interaction of two NADPH-dependent pathways. Within the tumor, increased lipid peroxidation and, consequently, saturation of the glutathione (GSH) system lead to the ferroptotic death of cancer cells. Systemically, redox imbalance and NADPH depletion impair corticosterone biosynthesis. Administration of dexamethasone, a potent glucocorticoid, increases food intake, normalizes glucose levels and utilization of nutritional substrates, delays cachexia onset, and extends the survival of tumor-bearing mice fed KD while preserving reduced tumor growth. Our study emphasizes the need to investigate the effects of systemic interventions on both the tumor and the host to accurately assess therapeutic potential. These findings may be relevant to clinical research efforts that investigate nutritional interventions such as KD in patients with cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetyl-L-Cystein, BioReagent, suitable for cell culture