Merck
  • Renal ischemia alters the transcriptomic and epigenetic profile of inflammatory genes in swine scattered tubular-like cells.
Renal ischemia alters the transcriptomic and epigenetic profile of inflammatory genes in swine scattered tubular-like cells.
Clinical science (London, England : 1979) (2023-08-22)
Sara Kazeminia, Xiang Y Zhu, Hui Tang, Kyra L Jordan, Ishran M Saadiq, Sandra M Herrmann, Alejandro R Chade, Maria V Irazabal, Lilach O Lerman, Alfonso Eirin
ZUSAMMENFASSUNG
Scattered tubular-like cells (STCs) are differentiated renal tubular cells that during recovery from ischemic injury dedifferentiate to repair other injured renal cells. Renal artery stenosis (RAS), often associated with chronic inflammatory injury, compromises the integrity and function of STCs, but the underlying mechanisms remain unknown. We hypothesized that RAS alters the transcriptomic and epigenetic profile of inflammatory genes in swine STCs. STCs were harvested from pig kidneys after 10 weeks of RAS or sham (n=6 each). STC mRNA profiles of inflammatory genes were analyzed using high-throughput mRNA-sequencing (seq) and their DNA methylation (5mC) and hydroxymethylation (5hmC) profiles by DNA immunoprecipitation and next-generation sequencing (MeDIP-seq) (n=3 each), followed by an integrated (mRNA-seq/MeDIP-seq) analysis. STC protein expression of candidate differentially expressed (DE) genes and common proinflammatory proteins were subsequently assessed in vitro before and after epigenetic (Bobcat339) modulation. mRNA-seq identified 57 inflammatory genes up-regulated in RAS-STCs versus Normal-STCs (>1.4 or <0.7-fold, P<0.05), of which 14% exhibited lower 5mC and 5% higher 5hmC levels in RAS-STCs versus Normal-STCs, respectively. Inflammatory gene and protein expression was higher in RAS-STCs compared with Normal-STCs but normalized after epigenetic modulation. These observations highlight a novel modulatory mechanism of this renal endogenous repair system and support development of epigenetic or anti-inflammatory therapies to preserve the reparative capacity of STCs in individuals with RAS.