Merck
  • The lyon clinical olfactory test: validation and measurement of hyposmia and anosmia in healthy and diseased populations.
The lyon clinical olfactory test: validation and measurement of hyposmia and anosmia in healthy and diseased populations.
International journal of otolaryngology (2011-11-03)
Catherine Rouby, Thierry Thomas-Danguin, Michel Vigouroux, Gabriela Ciuperca, Tao Jiang, Jérôme Alexanian, Mathieu Barges, Isabelle Gallice, Jean-Louis Degraix, Gilles Sicard
ZUSAMMENFASSUNG
The LCOT is a self-administered test designed to assess olfactory deficits. Altogether, 525 subjects contributed to the validation. Elderly participants were well represented in this sample. In a validation study (study 1), 407 healthy and 17 anosmic volunteers between 15 and 91 years of age underwent threshold, supraliminal detection, and identification testing. Cutoff values for normosmia and hyposmia were calculated and applied in a second study in a group of patients with smell complaints and in a group of Alzheimer patients with age-matched controls. Incidence of smell deficit was estimated at 5.6% in the healthy population of study 1, and at 16% in the elderly control group of study 2. Assessment of the ability of each subtest to discriminate between groups showed that LCOT is relevant to differentiating between perception and identification deficits and between Alzheimer's and hyposmic patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mineralöl, BioReagent, Molecular Biology, light oil
Sigma-Aldrich
Mineralöl, light oil, (neat)
Sigma-Aldrich
Mineralöl, light oil, suitable for mouse embryo cell culture, BioXtra
Sigma-Aldrich
Mineralöl, light
Sigma-Aldrich
Diethylphthalat, 99.5%
Sigma-Aldrich
Mineralöl, suitable for preparation of Nujol mulls for infrared spectroscopy, light oil
Sigma-Aldrich
Mineralöl, heavy
Sigma-Aldrich
Mineralöl, meets USP testing specifications
Sigma-Aldrich
Mineralöl, PCR Reagent
Sigma-Aldrich
Diethylphthalat, ≥99%
Sigma-Aldrich
Mineralöl, BioUltra, Molecular Biology
Supelco
Diethylphthalat, PESTANAL®, analytical standard