Merck
PAF1c links S-phase progression to immune evasion and MYC function in pancreatic carcinoma.
Nature communications (2024-02-17)
Abdallah Gaballa, Anneli Gebhardt-Wolf, Bastian Krenz, Greta Mattavelli, Mara John, Giacomo Cossa, Silvia Andreani, Christina Schülein-Völk, Francisco Montesinos, Raphael Vidal, Carolin Kastner, Carsten P Ade, Burkhard Kneitz, Georg Gasteiger, Peter Gallant, Mathias Rosenfeldt, Angela Riedel, Martin Eilers
ZUSAMMENFASSUNG
In pancreatic ductal adenocarcinoma (PDAC), endogenous MYC is required for S-phase progression and escape from immune surveillance. Here we show that MYC in PDAC cells is needed for the recruitment of the PAF1c transcription elongation complex to RNA polymerase and that depletion of CTR9, a PAF1c subunit, enables long-term survival of PDAC-bearing mice. PAF1c is largely dispensable for normal proliferation and regulation of MYC target genes. Instead, PAF1c limits DNA damage associated with S-phase progression by being essential for the expression of long genes involved in replication and DNA repair. Surprisingly, the survival benefit conferred by CTR9 depletion is not due to DNA damage, but to T-cell activation and restoration of immune surveillance. This is because CTR9 depletion releases RNA polymerase and elongation factors from the body of long genes and promotes the transcription of short genes, including MHC class I genes. The data argue that functionally distinct gene sets compete for elongation factors and directly link MYC-driven S-phase progression to tumor immune evasion.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Monoklonaler Anti-Vinculin-Antikörper in Maus hergestellte Antikörper, clone hVIN-1, ascites fluid
Sigma-Aldrich
Trypsin-Inhibitor aus Glycine max (soybean), powder, BioReagent, suitable for cell culture
Kaninchen-IgG, HRP-verknüpfter Vollantikörper, Cytiva NA934-1ML
Maus-IgG, HRP-verknüpfter Vollantikörper, Cytiva NA931-1ML