Merck
  • Brain Short-Chain Fatty Acids Induce ACSS2 to Ameliorate Depressive-Like Behavior via PPARγ-TPH2 Axis.
Brain Short-Chain Fatty Acids Induce ACSS2 to Ameliorate Depressive-Like Behavior via PPARγ-TPH2 Axis.
Research (Washington, D.C.) (2024-06-28)
Nuo Chen, Xinyi Xu, Yaxin Guo, Ming Zhao, Yubin Li, Tian Zhou, Xinyue Zhang, Jie Gao, Faliang Zhu, Chun Guo, Yongyu Shi, Qun Wang, Wenxian Wu, Lining Zhang, Yan Li
ZUSAMMENFASSUNG
Short-chain fatty acids (SCFAs) have been increasingly evidenced to be important bioactive metabolites of the gut microbiota and transducers in controlling diverse psychiatric or neurological disorders via the microbiota-gut-brain axis. However, the precise mechanism by which brain SCFAs extert multiple beneficial effects is not completely understood. Our previous research has demonstrated that the acetyl-coenzyme A synthetase short-chain family member 2 (ACSS2) is a novel target of the rapid and long-lasting antidepressant responses. Here, we show that micromolar SCFAs significantly augment both total cellular and nuclear ACSS2 to trigger tryptophan hydroxylase 2 (TPH2) promoter histone acetylation and its transcription in SH-SY5Y cells. In chronic-restraint-stress-induced depression mice, neuronal ACSS2 knockdown by stereotaxic injection of adeno-associated virus in the hippocampus abolished SCFA-mediated improvements in depressive-like behaviors of mice, supporting that ACSS2 is required for SCFA-mediated antidepressant responses. Mechanistically, the peroxisome-proliferator-activated receptor gamma (PPARγ) is identified as a novel partner of ACSS2 to activate TPH2 transcription. Importantly, PPARγ is also responsible for SCFA-mediated antidepressant-like effects via ACSS2-TPH2 axis. To further support brain SCFAs as a therapeutic target for antidepressant effects, d-mannose, which is a naturally present hexose, can significantly reverse the dysbiosis of gut microbiota in the chronic-restraint-stress-exposure mice and augment brain SCFAs to protect against the depressive-like behaviors via ACSS2-PPARγ-TPH2 axis. In summary, brain SCFAs can activate ACSS2-PPARγ-TPH2 axis to play the antidepressive-like effects, and d-mannose is suggested to be an inducer of brain SCFAs in resisting depression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ANTI-FLAG® in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Phenylmethylsulfonylfluorid, Phenylmethylsulfonyl Fluoride, CAS 329-98-6, is an irreversible inhibitor of serine proteases. It causes sulfonylation of the active-site serine residues.