Merck
AMPK-Nrf2 Signaling Pathway in Phrenic Motoneurons following Cervical Spinal Cord Injury.
Antioxidants (Basel, Switzerland) (2022-09-24)
Pauline Michel-Flutot, Laurie Efthimiadi, Lynda Djerbal, Therese B Deramaudt, Marcel Bonay, Stéphane Vinit
ZUSAMMENFASSUNG
High spinal cord injuries (SCI) induce the deafferentation of phrenic motoneurons, leading to permanent diaphragm paralysis. This involves secondary injury associated with pathologic and inflammatory processes at the site of injury, and at the level of phrenic motoneurons. In the present study, we evaluated the antioxidant response in phrenic motoneurons involving the AMPK-Nrf2 signaling pathway following C2 spinal cord lateral hemi-section in rats. We showed that there is an abrupt reduction in the expression of phosphorylated AMPK and Nrf2 at one hour post-injury in phrenic motoneurons. A rebound is then observed at one day post-injury, reflecting a return to homeostasis condition. In the total spinal cord around phrenic motoneurons, the increase in phosphorylated AMPK and Nrf2 occurred at three days post-injury, showing the differential antioxidant response between phrenic motoneurons and other cell types. Taken together, our results display the implication of the AMPK-Nrf2 signaling pathway in phrenic motoneurons' response to oxidative stress following high SCI. Harnessing this AMPK-Nrf2 signaling pathway could improve the antioxidant response and help in spinal rewiring to these deafferented phrenic motoneurons to improve diaphragm activity in patients suffering high SCI.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-GAPDH-Maus-mAb (6C5), liquid, clone 6C5, Calbiochem®
Sigma-Aldrich
Anti-Cholera Toxin, B-Subunit Goat pAb, lyophilized, Calbiochem®
Sigma-Aldrich
Anti-Makrophagen/Monozyten-Antikörper, Klon ED-1, clone ED-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αM [CD11b] Antibody, clone OX-42, clone OX-42, Chemicon®, from mouse
Sigma-Aldrich
Anti-Nitric Oxide Synthase II Antibody, serum, Chemicon®