Merck
  • Intracellular free calcium mediates glioma cell detachment and cytotoxicity after photodynamic therapy.
Intracellular free calcium mediates glioma cell detachment and cytotoxicity after photodynamic therapy.
Lasers in medical science (2009-02-10)
Xin Hong, Feng Jiang, Steven N Kalkanis, Zheng Gang Zhang, Xuepeng Zhang, Xuguang Zheng, Hao Jiang, Michael Chopp
ZUSAMMENFASSUNG
Photofrin photodynamic therapy (PDT) caused a dose-dependent decrease of enzymatic cell detachment by trypsin/ethylenediamine tetra-acetic acid (EDTA) in human glioma U251n and U87 cells. This happened coincidently with the increase of intracellular free calcium ([Ca(2+)](i)). Thapsigargin, which increased [Ca(2+)](i), induced further decrease in enzymatic cell detachment and increased cytotoxicity. Opposite effects were observed when 1,2-bis(2-aminophenoxy) ethane-N,N,N',N'-tetra-acetic acid tetrakis, an intracellular Ca(2+) chelator, was used. PDT-induced changes in [Ca(2+)](i) and cell detachment were not blocked by calcium channel antagonists nickel (Ni(2+)) or nimodipine, nor were they altered when cells were irradiated in a buffer free from Ca(2+) and magnesium (Mg(2+)), suggesting that [Ca(2+)](i) is derived from the internal calcium stores. Decreased cell migration was observed after PDT, as assessed by chemotactic and wound-healing assays. Our findings indicated that internal calcium store-derived [Ca(2+)](i) plays an important role in PDT-induced enzymatic cell detachment decrease and cytotoxicity. Cell migration may be affected by these changes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Digitonin, Used as non-ionic detergent
Sigma-Aldrich
Digitonin, ~50% (TLC)
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)