Merck
Mechanism and specificity of pentachloropseudilin-mediated inhibition of myosin motor activity.
The Journal of biological chemistry (2011-06-18)
Krishna Chinthalapudi, Manuel H Taft, René Martin, Sarah M Heissler, Matthias Preller, Falk K Hartmann, Hemma Brandstaetter, John Kendrick-Jones, Georgios Tsiavaliaris, Herwig O Gutzeit, Roman Fedorov, Folma Buss, Hans-Joachim Knölker, Lynne M Coluccio, Dietmar J Manstein
ZUSAMMENFASSUNG
Here, we report that the natural compound pentachloropseudilin (PClP) acts as a reversible and allosteric inhibitor of myosin ATPase and motor activity. IC(50) values are in the range from 1 to 5 μm for mammalian class-1 myosins and greater than 90 μm for class-2 and class-5 myosins, and no inhibition was observed with class-6 and class-7 myosins. We show that in mammalian cells, PClP selectively inhibits myosin-1c function. To elucidate the structural basis for PClP-induced allosteric coupling and isoform-specific differences in the inhibitory potency of the compound, we used a multifaceted approach combining direct functional, crystallographic, and in silico modeling studies. Our results indicate that allosteric inhibition by PClP is mediated by the combined effects of global changes in protein dynamics and direct communication between the catalytic and allosteric sites via a cascade of small conformational changes along a conserved communication pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pentachloropseudilin, ≥98% (HPLC)