Merck
  • Transcriptional dynamics in the protozoan parasite Sarcocystis neurona and mammalian host cells after treatment with a specific inhibitor of apicomplexan mRNA polyadenylation
Transcriptional dynamics in the protozoan parasite Sarcocystis neurona and mammalian host cells after treatment with a specific inhibitor of apicomplexan mRNA polyadenylation
PLoS ONE (2021)