Merck
  • The Effects of Platelet-Derived Growth Factor-BB on Bone Marrow Stromal Cell-Mediated Vascularized Bone Regeneration.
The Effects of Platelet-Derived Growth Factor-BB on Bone Marrow Stromal Cell-Mediated Vascularized Bone Regeneration.
Stem cells international (2018-12-06)
Maolin Zhang, Wenwen Yu, Kunimichi Niibe, Wenjie Zhang, Hiroshi Egusa, Tingting Tang, Xinquan Jiang
ZUSAMMENFASSUNG
Regenerative medicine for bone tissue mainly depends on efficient recruitment of endogenous or transplanted stem cells to guide bone regeneration. Platelet-derived growth factor (PDGF) is a functional factor that has been widely used in tissue regeneration and repair. However, the short half-life of PDGF limits its efficacy, and the mechanism by which PDGF regulates stem cell-based bone regeneration still needs to be elucidated. In this study, we established genetically modified PDGF-B-overexpressing bone marrow stromal cells (BMSCs) using a lentiviral vector and then explored the mechanism by which PDGF-BB regulates BMSC-based vascularized bone regeneration. Our results demonstrated that PDGF-BB increased osteogenic differentiation but inhibited adipogenic differentiation of BMSCs via the extracellular signal-related kinase 1/2 (ERK1/2) signaling pathway. In addition, secreted PDGF-BB significantly enhanced human umbilical vein endothelial cell (HUVEC) migration and angiogenesis via the phosphatidylinositol 3 kinase (PI3K)/AKT and ERK1/2 signaling pathways. We evaluated the effect of PDGF-B-modified BMSCs on bone regeneration using a critical-sized rat calvarial defect model. Radiography, micro-CT, and histological analyses revealed that PDGF-BB overexpression improved BMSC-mediated angiogenesis and osteogenesis during bone regeneration. These results suggest that PDGF-BB facilitates BMSC-based bone regeneration by enhancing the osteogenic and angiogenic abilities of BMSCs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
TissueFab® PODS Growth Factor Loaded Bioink Kit, Bone differentiation