Merck
  • Lotilaner Ophthalmic Solution 0.25% for Demodex Blepharitis: Randomized, Vehicle-Controlled, Multicenter, Phase 3 Trial (Saturn-2).
Lotilaner Ophthalmic Solution 0.25% for Demodex Blepharitis: Randomized, Vehicle-Controlled, Multicenter, Phase 3 Trial (Saturn-2).
Ophthalmology (2023-06-08)
Ian Benjamin Gaddie, Eric D Donnenfeld, Paul Karpecki, Patrick Vollmer, Gregg J Berdy, Jared D Peterson, Blake Simmons, Aimée R P Edell, William E Whitson, Joseph B Ciolino, Stephanie N Baba, Mark Holdbrook, José Trevejo, John Meyer, Elizabeth Yeu
ZUSAMMENFASSUNG
To evaluate the safety and efficacy of lotilaner ophthalmic solution 0.25% compared with vehicle for the treatment of Demodex blepharitis. Prospective, randomized, double-masked, vehicle-controlled, multicenter, phase 3 clinical trial. Four hundred twelve patients with Demodex blepharitis were assigned randomly in a 1:1 ratio to receive either lotilaner ophthalmic solution 0.25% (study group) or vehicle without lotilaner (control group). Patients with Demodex blepharitis treated at 21 United States clinical sites were assigned either to the study group (n = 203) to receive lotilaner ophthalmic solution 0.25% or to the control group (n = 209) to receive vehicle without lotilaner bilaterally twice daily for 6 weeks. Collarettes and erythema were graded for each eyelid at screening and at all visits after baseline. At screening and on days 15, 22, and 43, 4 or more eyelashes were epilated from each eye, and the number of Demodex mites present on the lashes was counted with a microscope. Mite density was calculated as the number of mites per lash. Outcome measures included collarette cure (collarette grade 0), clinically meaningful collarette reduction to 10 collarettes or fewer (grade 0 or 1), mite eradication (0 mites/lash), erythema cure (grade 0), composite cure (grade 0 for collarettes as well as erythema), compliance with the drop regimen, drop comfort, and adverse events. At day 43, the study group achieved a statistically significant (P < 0.0001) higher proportion of patients with collarette cure (56.0% vs. 12.5%), clinically meaningful collarette reduction to 10 collarettes or fewer (89.1% vs. 33.0%), mite eradication (51.8% vs. 14.6%), erythema cure (31.1% vs. 9.0%), and composite cure (19.2% vs. 4.0%) than the control group. High compliance with the drop regimen (mean ± standard deviation, 98.7 ± 5.3%) in the study group was observed, and 90.7% of patients found the drops to be neutral to very comfortable. Twice-daily treatment with lotilaner ophthalmic solution 0.25% for 6 weeks generally was safe and well tolerated and met the primary end point and all secondary end points for the treatment of Demodex blepharitis compared with vehicle control. Proprietary or commercial disclosure may be found in the Footnotes and Disclosures at the end of this article.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Lotilaner, ≥98% (HPLC)