Merck
Polysaccharide hydrogel based 3D printed tumor models for chemotherapeutic drug screening.
Scientific reports (2021-01-13)
Aragaw Gebeyehu, Sunil Kumar Surapaneni, John Huang, Arindam Mondal, Vivian Ziwen Wang, Nana Fatima Haruna, Arvind Bagde, Peggy Arthur, Shallu Kutlehria, Nil Patel, Arun K Rishi, Mandip Singh
ZUSAMMENFASSUNG
A series of stable and ready-to-use bioinks have been developed based on the xeno-free and tunable hydrogel (VitroGel) system. Cell laden scaffold fabrication with optimized polysaccharide-based inks demonstrated that Ink H4 and RGD modified Ink H4-RGD had excellent rheological properties. Both bioinks were printable with 25-40 kPa extrusion pressure, showed 90% cell viability, shear-thinning and rapid shear recovery properties making them feasible for extrusion bioprinting without UV curing or temperature adjustment. Ink H4-RGD showed printability between 20 and 37 °C and the scaffolds remained stable for 15 days at temperature of 37 °C. 3D printed non-small-cell lung cancer (NSCLC) patient derived xenograft cells (PDCs) showed rapid spheroid growth of size around 500 µm in diameter and tumor microenvironment formation within 7 days. IC50 values demonstrated higher resistance of 3D spheroids to docetaxel (DTX), doxorubicin (DOX) and erlotinib compared to 2D monolayers of NSCLC-PDX, wild type triple negative breast cancer (MDA-MB-231 WT) and lung adenocarcinoma (HCC-827) cells. Results of flow property, shape fidelity, scaffold stability and biocompatibility of H4-RGD suggest that this hydrogel could be considered for 3D cell bioprinting and also for in-vitro tumor microenvironment development for high throughput screening of various anti-cancer drugs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
TissueFab® Bioink Mixing Kit, 10:1 Mixing component pack
TissueFab® xeno-free bioink, IKVAV modified, facile curable
TissueFab® xeno-free bioink, (YIGSR modified), (facile curable)