Merck
A multi-marker assay to distinguish malignant melanomas from benign nevi.
Proceedings of the National Academy of Sciences of the United States of America (2009-04-01)
Mohammed Kashani-Sabet, Javier Rangel, Sima Torabian, Mehdi Nosrati, Jeff Simko, David M Jablons, Dan H Moore, Chris Haqq, James R Miller, Richard W Sagebiel
ZUSAMMENFASSUNG
The histopathological diagnosis of melanoma can be challenging. No currently used molecular markers accurately distinguish between nevus and melanoma. Recent transcriptome analyses have shown the differential expression of several genes in melanoma progression. Here, we describe a multi-marker diagnostic assay using 5 markers (ARPC2, FN1, RGS1, SPP1, and WNT2) overexpressed in melanomas. Immunohistochemical marker expression was analyzed in 693 melanocytic neoplasms comprising a training set (tissue microarray of 534 melanomas and nevi), and 4 independent validation sets: tissue sections of melanoma arising in a nevus; dysplastic nevi; Spitz nevi; and misdiagnosed melanocytic neoplasms. Both intensity and pattern of expression were scored for each marker. Based on the differential expression of these 5 markers between nevi and melanomas in the training set, a diagnostic algorithm was obtained. Using this algorithm, the lesions in the validation sets were diagnosed as nevus or melanoma, and the results were compared with the known histological diagnoses. Both the intensity and pattern of expression of each marker were significantly different in melanomas compared to nevi. The diagnostic algorithm exploiting these differences achieved a specificity of 95% and a sensitivity of 91% in the training set. In the validation sets, the multi-marker assay correctly diagnosed a high percentage of melanomas arising in a nevus, Spitz nevi, dysplastic nevi, and misdiagnosed lesions. The multi-marker assay described here can aid in the diagnosis of melanoma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Wasserstoffperoxid -Lösung, tested according to Ph. Eur.