Merck
  • Imidazole metalloporphyrins as photosensitizers for photodynamic therapy: role of molecular charge, central metal and hydroxyl radical production.
Imidazole metalloporphyrins as photosensitizers for photodynamic therapy: role of molecular charge, central metal and hydroxyl radical production.
Cancer letters (2009-04-07)
Pawel Mroz, Jayeeta Bhaumik, Dilek K Dogutan, Zarmeneh Aly, Zahra Kamal, Laiqua Khalid, Hooi Ling Kee, David F Bocian, Dewey Holten, Jonathan S Lindsey, Michael R Hamblin
ZUSAMMENFASSUNG
The in vitro photodynamic therapy activity of four imidazole-substituted metalloporphyrins has been studied using human (HeLa) and mouse (CT26) cancer cell lines: an anionic Zn porphyrin and a homologous series of three cationic Zn, Pd or InCl porphyrins. A dramatic difference in phototoxicity was found: Pd cationic>InCl cationic>Zn cationic>Zn anionic. HeLa cells were more susceptible than CT26 cells. Induction of apoptosis was demonstrated using a fluorescent caspase assay. The anionic Zn porphyrin localized in lysosomes while the cationic Zn porphyrin localized in lysosomes and mitochondria, as assessed by fluorescence microscopy. Studies using fluorescent probes suggested that the cationic Pd porphyrin produced more hydroxyl radicals as the reactive oxygen species. Thus, the cationic Pd porphyrin has high potential as a photosensitizer and gives insights into characteristics for improved molecular designs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trypsin-EDTA-Lösung, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
Trypsin-EDTA-Lösung, 1 ×, sterile; sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
Trypsin-EDTA-Lösung, 10 ×, sterile-filtered, BioReagent, suitable for cell culture, 5.0 g porcine trypsin and 2 g EDTA, 4Na per liter of 0.9% sodium chloride
Sigma-Aldrich
Trypanblau, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Rhodamin 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Trypanblau, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypsin-EDTA-Lösung, 1 ×, sterile-filtered, BioReagent, suitable for cell culture, 500 BAEE units porcine trypsin and 180 μg EDTA, 4Na per ml in Dulbecco′s PBS without calcium and magnesium
Sigma-Aldrich
Rhodamin 123, BioReagent, suitable for fluorescence, ≥85% (HPLC)