Merck
  • The meningococcal ABC-Type L-glutamate transporter GltT is necessary for the development of experimental meningitis in mice.
The meningococcal ABC-Type L-glutamate transporter GltT is necessary for the development of experimental meningitis in mice.
Infection and immunity (2009-06-17)
Roberta Colicchio, Susanna Ricci, Florentia Lamberti, Caterina Pagliarulo, Chiara Pagliuca, Velia Braione, Tiziana Braccini, Adelfia Talà, Donatella Montanaro, Sergio Tripodi, Marcella Cintorino, Giancarlo Troncone, Cecilia Bucci, Gianni Pozzi, Carmelo B Bruni, Pietro Alifano, Paola Salvatore
ZUSAMMENFASSUNG
Experimental animal models of bacterial meningitis are useful to study the host-pathogen interactions occurring at the cerebral level and to analyze the pathogenetic mechanisms behind this life-threatening disease. In this study, we have developed a mouse model of meningococcal meningitis based on the intracisternal inoculation of bacteria. Experiments were performed with mouse-passaged serogroup C Neisseria meningitidis. Survival and clinical parameters of infected mice and microbiological and histological analysis of the brain demonstrated the establishment of meningitis with features comparable to those of the disease in humans. When using low bacterial inocula, meningococcal replication in the brain was very efficient, with a 1,000-fold increase of viable counts in 18 h. Meningococci were also found in the blood, spleens, and livers of infected mice, and bacterial loads in different organs were dependent on the infectious dose. As glutamate uptake from the host has been implicated in meningococcal virulence, mice were infected intracisternally with an isogenic strain deficient in the ABC-type L-glutamate transporter GltT. Noticeably, the mutant was attenuated in virulence in mixed infections, indicating that wild-type bacteria outcompeted the GltT-deficient meningococci. The data show that the GltT transporter plays a role in meningitis and concomitant systemic infection, suggesting that meningococci may use L-glutamate as a nutrient source and as a precursor to synthesize the antioxidant glutathione.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saponin, Molecular Biology, used as non-ionic surfactant
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Saponin, used as non-ionic surfactant
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.