Merck
  • Multiple mild heat-shocks decrease the Gompertz component of mortality in Caenorhabditis elegans.
Multiple mild heat-shocks decrease the Gompertz component of mortality in Caenorhabditis elegans.
Experimental gerontology (2009-07-08)
Deqing Wu, James R Cypser, Anatoli I Yashin, Thomas E Johnson
ZUSAMMENFASSUNG
Exposure to mild heat-stress (heat-shock) can significantly increase the life expectancy of the nematode Caenorhabditis elegans. A single heat-shock early in life extends longevity by 20% or more and affects life-long mortality by decreasing initial mortality only; the rate of increase in subsequent mortality (Gompertz component) is unchanged. Repeated mild heat-shocks throughout life have a larger effect on life span than does a single heat-shock early in life. Here, we ask how multiple heat-shocks affect the mortality trajectory in nematodes and find increases of life expectancy of close to 50% and of maximum longevity as well. We examined mortality using large numbers of animals and found that multiple heat-shocks not only decrease initial mortality, but also slow the Gompertz rate of increase in mortality. Thus, multiple heat-shocks have anti-aging hormetic effects and represent an effective approach for modulating aging.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5-Fluor-2′-Desoxyuridin, thymidylate synthase inhibitor