Merck
Serotonin targets inhibitory synapses to induce modulation of network functions.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences (2009-08-05)
Till Manzke, Mathias Dutschmann, Gerald Schlaf, Michael Mörschel, Uwe R Koch, Evgeni Ponimaskin, Olivier Bidon, Peter M Lalley, Diethelm W Richter
ZUSAMMENFASSUNG
The cellular effects of serotonin (5-HT), a neuromodulator with widespread influences in the central nervous system, have been investigated. Despite detailed knowledge about the molecular biology of cellular signalling, it is not possible to anticipate the responses of neuronal networks to a global action of 5-HT. Heterogeneous expression of various subtypes of serotonin receptors (5-HTR) in a variety of neurons differently equipped with cell-specific transmitter receptors and ion channel assemblies can provoke diverse cellular reactions resulting in various forms of network adjustment and, hence, motor behaviour. Using the respiratory network as a model for reciprocal synaptic inhibition, we demonstrate that 5-HT(1A)R modulation primarily affects inhibition through glycinergic synapses. Potentiation of glycinergic inhibition of both excitatory and inhibitory neurons induces a functional reorganization of the network leading to a characteristic change of motor output. The changes in network operation are robust and help to overcome opiate-induced respiratory depression. Hence, 5-HT(1A)R activation stabilizes the rhythmicity of breathing during opiate medication of pain.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,3′-Diaminobenzidin, 97%
Sigma-Aldrich
Polyethylenglycol-Lösung, Hybri-Max, 50 % (w/v), average mol wt 1,450, 0.2 μm filtered, BioReagent, suitable for hybridoma
Sigma-Aldrich
Polyethylenglykol -Lösung, 40 % (w/w) in H2O, average mol wt 8,000
Sigma-Aldrich
3,3′-Diaminobenzidin, 97% (HPLC)
Sigma-Aldrich
Polyethylenglykol -Lösung, BioUltra, Molecular Biology, 1,000, ~50% in H2O
Sigma-Aldrich
Strychnine hydrochloride
Sigma-Aldrich
Polyethylenglycol-/Dimethylsulfoxid-Lösung, Hybri-Max, average mol wt 1,450, 50 % (w/v), 0.2 μm filtered, BioReagent, suitable for hybridoma
Sigma-Aldrich
Anti-Substance P Receptor Antibody, Chemicon®, from guinea pig