Merck
  • Critical role of the interleukin-17/interleukin-17 receptor axis in regulating host susceptibility to respiratory infection with Chlamydia species.
Critical role of the interleukin-17/interleukin-17 receptor axis in regulating host susceptibility to respiratory infection with Chlamydia species.
Infection and immunity (2009-09-10)
Xiaohui Zhou, Qiangwei Chen, Jessica Moore, Jay K Kolls, Scott Halperin, Jun Wang
ZUSAMMENFASSUNG
The specific contribution of interleukin-17/interleukin-17 receptor (IL-17/IL-17R)-mediated responses in regulating host susceptibility against obligatory intracellular Chlamydia infection was investigated in C57BL/6 and C3H/HeN mice during Chlamydia muridarum respiratory infection. We demonstrated that Chlamydia stimulated IL-17/IL-17R-associated responses in both Chlamydia-resistant C57BL/6 and Chlamydia-susceptible C3H/HeN mice. However, C3H/HeN mice developed a significantly greater IL-17/IL-17R-associated response than C57BL/6 mice did. This was reflected by an increase in IL-17 mRNA expression, a higher recall IL-17 production from splenocytes upon antigen restimulation, and higher production of Th17-related cytokines (IL-23 and IL-6) and chemokines (chemokine [C-X-C motif] ligand 2 [CXCL1]/keratinocyte-derived chemokine [KC] and CXCL2/macrophage inflammatory protein 1 [MIP2]) in C3H/HeN mice than in C57BL/6 mice. Furthermore, C3H/HeN mice displayed a massive accumulation of activated and preactivated neutrophils in the airway and lung parenchyma compared to their C57BL/6 counterparts. We further demonstrated that the skewed IL-17/Th17 profile in C3H/HeN mice was predisposed by a higher basal level of IL-17 receptor C (IL-17RC) expression and then further amplified by a higher inducible IL-17RA expression in lungs. Most importantly, in vivo delivery of IL-17RA antagonist that resulted in a 50% reduction in the neutrophilic infiltration in lungs was able to reverse the susceptible phenotype of C3H/HeN mice to respiratory Chlamydia infection. Thus, our data for the first time have demonstrated a critical role for the IL-17/IL-17R axis in regulating host susceptibility to Chlamydia infection in mice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Hexadecyltrimethylammoniumchlorid -Lösung, 25 wt. % in H2O
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Saccharose, meets USP testing specifications
Sigma-Aldrich
4-Chlor-1-Naphthol, crystalline
Sigma-Aldrich
4-Chlor-1-Naphthol, tablet
Supelco
Saccharose, analytical standard, for enzymatic assay kit SCA20