Merck
  • Peptidoglycan from lactobacilli inhibits interleukin-12 production by macrophages induced by Lactobacillus casei through Toll-like receptor 2-dependent and independent mechanisms.
Peptidoglycan from lactobacilli inhibits interleukin-12 production by macrophages induced by Lactobacillus casei through Toll-like receptor 2-dependent and independent mechanisms.
Immunology (2009-09-25)
Kan Shida, Junko Kiyoshima-Shibata, Rumi Kaji, Masato Nagaoka, Masanobu Nanno
ZUSAMMENFASSUNG
We previously showed that Lactobacillus strains having a rigid cell wall resistant to intracellular digestion can stimulate macrophages to induce large a quantity of interleukin-12 (IL-12). In this study, we examined the influence of lactobacilli and bacterial cell wall components on IL-12 production by macrophages that was induced by Lactobacillus casei, which has a rigid cell wall. Easily digestible lactobacilli such as Lactobacillus johnsonii and Lactobacillus plantarum or their intact cell walls (ICWs) weakly or very weakly induced IL-12 production by macrophages, and inhibitedL. casei-induced IL-12 production. While the ICW of L. casei was resistant to intracellular digestion and did not inhibit L. casei-induced IL-12 production, its polysaccharide-depleted ICW, i.e. intact peptidoglycan, was sensitive to intracellular digestion and inhibited L. casei-induced IL-12 production. Furthermore, the peptidoglycans of L. johnsonii, L. plantarum and Staphylococcus aureus also inhibited L. casei-induced IL-12 production. Peptidoglycans from lactobacilli suppressed L. casei-induced expression of IL-12p40 but not IL-12p35 mRNA. Inhibition of IL-12 production by peptidoglycan was mitigated in Toll-like receptor 2 (TLR2)-deficient macrophages compared with the inhibition in wild-type macrophages. A derivative of the minimal structural unit of peptidoglycan (6-O-stearoyl-muramyl dipeptide) recognized by nucleotide-binding oligomerization domain 2 (NOD2) could also suppress L. casei-induced IL-12 production. These findings demonstrate that easily digestible bacteria and peptidoglycan suppress IL-12 production through pattern recognition receptors such as TLR2 and NOD2. IL-12 production in the gut may be negatively regulated by the simultaneous inhibitory actions of various resident bacteria that are susceptible to intracellular digestion.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
RPMI-1640-Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640-Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640-Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640-Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cytochalasin D aus Zygosporium masonii, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
RPMI-1640-Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640-Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640-Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640-Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640-Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Giemsa-Färbung, certified by the BSC
Sigma-Aldrich
Cytochalasin D aus Zygosporium masonii, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Sigma-Aldrich
RPMI-1640-Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640-Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
SAFC
RPMI-1640-Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640-Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640-Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications
SAFC
RPMI-1640-Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Giemsa-Färbung, technical grade, used as a blood stain
Sigma-Aldrich
RPMI-1640-Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture