- Caged Q-rhodamine dextran: a new photoactivated fluorescent tracer.
Caged Q-rhodamine dextran: a new photoactivated fluorescent tracer.
Bioorganic & medicinal chemistry letters (2001-08-22)
K R Gee, E S Weinberg, D J Kozlowski
PMID11514165
ZUSAMMENFASSUNG
An amine-reactive caged rhodamine was synthesized and conjugated to aminodextran. The resulting tracer was injected into a single cell zebrafish embryo, and a portion of the tracer was photolyzed in a single cell after development. The resulting fluorescent cell was imaged by fluorescence microscopy through a single round of cell division.