Merck
  • Pulmonary gene delivery of hybrid vector, lipopolyplex containing N-lauroylsarcosine, via the systemic route.
Pulmonary gene delivery of hybrid vector, lipopolyplex containing N-lauroylsarcosine, via the systemic route.
J. Controlled Release (2009)
Kurosaki T, Kishikawa R, et al.