Merck
  • Effect of water-soluble polymers on the physical stability of aqueous polymeric dispersions and their implications on the drug release from coated pellets.
Effect of water-soluble polymers on the physical stability of aqueous polymeric dispersions and their implications on the drug release from coated pellets.
Drug Devel. Ind. Pharm. (2010)
Dashevsky A, Ahmed AR, et al.