Merck
  • 4-Vinylcyclohexene diepoxide (VCD) inhibits mammary epithelial differentiation and induces fibroadenoma formation in female Sprague Dawley rats.
4-Vinylcyclohexene diepoxide (VCD) inhibits mammary epithelial differentiation and induces fibroadenoma formation in female Sprague Dawley rats.
Reproductive toxicology (Elmsford, N.Y.) (2011-05-31)
Laura E Wright, Jennifer B Frye, Ashley L Lukefahr, Samuel L Marion, Patricia B Hoyer, David G Besselsen, Janet L Funk
ZUSAMMENFASSUNG
4-Vinylcyclohexene diepoxide (VCD), an occupational chemical that targets ovarian follicles and accelerates ovarian failure in rodents, was used to test the effect of early-onset reproductive senescence on mammary fibroadenoma formation. One-month female Sprague Dawley rats were dosed with VCD (80 mg/kg or 160 mg/kg) and monitored for 22 months for persistent estrus and tumor development. Only high-dose VCD treatment accelerated the onset of persistent estrus relative to controls. However, both doses of VCD accelerated mammary tumor onset by 5 months, increasing incidence to 84% (vs. 38% in controls). Tumor development was independent of time in persistent estrus, 17 β-estradiol, androstenedione and prolactin. Delay in VCD administration until after completion of mammary epithelial differentiation (3 months) did not alter tumor formation despite acceleration of ovarian senescence. VCD administration to 1-month rats acutely decreased mammary alveolar bud number and expression of β-casein, suggesting that VCD's tumorigenic effect requires exposure during mammary epithelial differentiation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Vinyl-cyclohexen-dioxid, purum, suitable for electron microscopy, mixture of isomers, ≥96.0% (GC)