Merck
  • Cocoa polyphenols prevent inflammation in the colon of azoxymethane-treated rats and in TNF-α-stimulated Caco-2 cells.
Cocoa polyphenols prevent inflammation in the colon of azoxymethane-treated rats and in TNF-α-stimulated Caco-2 cells.
The British Journal of Nutrition (2012)
Rodriguez-Ramiro I, Ramos S, et al.