Merck
  • Co-transcription of the gastrin and cholecystokinin genes with selective translation of gastrin mRNA in a human gastric carcinoma cell line.
Co-transcription of the gastrin and cholecystokinin genes with selective translation of gastrin mRNA in a human gastric carcinoma cell line.
FEBS letters (1992-08-31)
W W van Solinge, J F Rehfeld
ZUSAMMENFASSUNG
So far, no cells have been found to synthesize both of the homologous hormones, cholecystokinin and gastrin. Northern analysis and reverse transcription PCR showed, that the human gastric carcinoma cell line (AGS) expresses both a gastrin mRNA of 0.7 kb and a cholecystokinin transcript of 0.8 kb. A library of sequence-specific radioimmunoassays, cleavage with processing-like enzymes and chromatography subsequently revealed that the gastrin mRNA was translated into progastrin that was constitutively secreted into the medium (45 +/- 3 pmol/l). Neither procholecystokinin nor any of its processing products were detectable in cells and media. The results suggest that differentiation into gastrin- or cholecystokinin-producing cells may be regulated at the translational level. The gastric cell line, AGS, provides a model for studies of translational regulation of cell differentiation.