Merck
  • Immunocytochemical localization of the alpha 7 subunit of the nicotinic acetylcholine receptor in the rat central nervous system.
Immunocytochemical localization of the alpha 7 subunit of the nicotinic acetylcholine receptor in the rat central nervous system.
The Journal of comparative neurology (1994-11-15)
E Dominguez del Toro, J M Juiz, X Peng, J Lindstrom, M Criado
ZUSAMMENFASSUNG
Previous molecular cloning studies have revealed that alpha-bungarotoxin binding proteins present in the brain are members of the neuronal nicotinic acetylcholine receptor gene family. The alpha 7 subunit is structurally related to the agonist binding subunits present in the central and peripheral nervous systems and, when expressed in Xenopus oocytes, forms functional channels blockable by alpha-bungarotoxin. In the present study, three different monoclonal antibodies raised against the alpha 7 subunit were used to map its distribution throughout the central nervous system of the rat. Immunohistochemical localization revealed that the alpha 7 subunit is expressed in most regions of the brain, being, overall, well correlated with previous "in situ" localization of alpha 7 transcripts and alpha-bungarotoxin autoradiographic binding studies. Particularly strong immunoreactivity was observed in several sensory and motor nuclei of the brainstem as well as the red nucleus. At the cellular level, alpha 7 immunostaining was usually found both in somata and dendrites, whereas axonal and terminal labeling was not observed. The widespread distribution of the alpha 7 subunit polypeptide is consistent with immunoprecipitation data demonstrating that it is a component of the predominant subtype of brain alpha-bungarotoxin-sensitive nicotinic receptors.