- A cytokine-responsive IkappaB kinase that activates the transcription factor NF-kappaB.
A cytokine-responsive IkappaB kinase that activates the transcription factor NF-kappaB.
Nature (1997-08-07)
J A DiDonato, M Hayakawa, D M Rothwarf, E Zandi, M Karin
PMID9252186
ZUSAMMENFASSUNG
Nuclear transcription factors of the NF-kappaB/Rel family are inhibited by IkappaB proteins, which inactivate NF-kappaB by trapping it in the cell cytoplasm. Phosphorylation of IkappaBs marks them out for destruction, thereby relieving their inhibitory effect on NF-kappaB. A cytokine-activated protein kinase complex, IKK (for IkappaB kinase), has now been purified that phosphorylates IkappaBs on the sites that trigger their degradation. A component of IKK was molecularly cloned and identified as a serine kinase. IKK turns out to be the long-sought-after protein kinase that mediates the critical regulatory step in NF-kappaB activation.