Merck
  • Differential labelling of human sub-cellular compartments with fluorescent dye esters and expansion microscopy.
Differential labelling of human sub-cellular compartments with fluorescent dye esters and expansion microscopy.
Nanoscale (2023-11-09)
Thomas M D Sheard, Tayla B Shakespeare, Rajpinder S Seehra, Michael E Spencer, Kin M Suen, Izzy Jayasinghe
ZUSAMMENFASSUNG
Amine-reactive esters of aromatic fluorescent dyes are emerging as imaging probes for nondescript staining of cellular and tissue architectures. We characterised the staining patterns of 14 fluorescent dye ester species with varying physical and spectral properties in the broadly studied human HeLa cell line. When combined with the super-resolution technique expansion microscopy (ExM) involving swellable acrylamide hydrogels, fluorescent esters reveal nanoscale features including cytoplasmic membrane-bound compartments and nucleolar densities. We observe differential labelling patterns linked to the biochemical properties of the conjugated dye. Alterations in staining density and compartment specificity were seen depending on the timepoint of application in the ExM protocol. Additional complexity in labelling patterns was detected arising from inter-ester interactions. Our findings raise a number of considerations for the use of fluorescent esters. We demonstrate esters as a useful addition to the repertoire of stains of the cellular proteome, whether applied either on their own to visualise overall cellular morphology, or as counterstains providing ultrastructural context alongside specific target markers like antibodies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
BODIPY 630/650 X NHS ester, ≥85% (HPLC)