Merck
  • Apoptosis of Kinetin Riboside in Colorectal Cancer Cells Occurs by Promoting β-Catenin Degradation.
Apoptosis of Kinetin Riboside in Colorectal Cancer Cells Occurs by Promoting β-Catenin Degradation.
Journal of microbiology and biotechnology (2023-07-19)
TaeKyung Nam, Wonku Kang, Sangtaek Oh
ZUSAMMENFASSUNG
Kinetin riboside is a naturally produced cytokinin that displays strong antiproliferative activity in various human cancer cells. However, the mechanism of chemoprevention in colorectal cancer cells has not been elucidated. We used a cell-based reporter system to identify kinetin riboside as an antagonist of the Wnt/β-catenin pathway, which is aberrantly upregulated in colorectal cancer. Kinetin riboside suppressed β-catenin response transcription (CRT) by accelerating the degradation of intracellular β-catenin via a proteasomal degradation pathway. Pharmacological inhibition of glycogen synthase kinase-3β did not affect CRT downregulation. Kinetin riboside decreased the intracellular β-catenin levels in colorectal cancer cells with mutations in adenomatous polyposis coli (APC) and β-catenin. Consistently, kinetin riboside repressed expression of c-Myc and cyclin D1, β-catenin/T-cell factor (TCF)-dependent genes, and inhibited the proliferation of colorectal cancer cells. In addition, kinetin riboside stimulated apoptosis, as measured by an increase in annexin V-FITC-stained cells. These findings suggest that kinetin riboside exerts its anti-cancer activity by promoting β-catenin degradation and has significant potential as a chemopreventive agent for colorectal cancer cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
N6-Furfuryladenosine, ≥98% (HPLC)