Merck
  • Preclinical evaluation of stereopure antisense oligonucleotides for allele-selective lowering of mutant HTT.
Preclinical evaluation of stereopure antisense oligonucleotides for allele-selective lowering of mutant HTT.
Molecular therapy. Nucleic acids (2024-07-19)
Naoki Iwamoto, Yuanjing Liu, Maria Frank-Kamenetsky, Abbie Maguire, Wei Chou Tseng, Kristin Taborn, Nayantara Kothari, Ali Akhtar, Keith Bowman, Juili Dilip Shelke, Anthony Lamattina, Xiao Shelley Hu, Hyun Gyung Jang, Pachamuthu Kandasamy, Fangjun Liu, Ken Longo, Richard Looby, Meena, Jake Metterville, Qianli Pan, Erin Purcell-Estabrook, Mamoru Shimizu, Priyanka Shiva Prakasha, Stephany Standley, Hansini Upadhyay, Hailin Yang, Yuan Yin, Anderson Zhao, Christopher Francis, Mike Byrne, Elena Dale, Gregory L Verdine, Chandra Vargeese
ZUSAMMENFASSUNG
Huntington's disease (HD) is an autosomal dominant disease caused by the expansion of cytosine-adenine-guanine (CAG) repeats in one copy of the HTT gene (mutant HTT, mHTT). The unaffected HTT gene encodes wild-type HTT (wtHTT) protein, which supports processes important for the health and function of the central nervous system. Selective lowering of mHTT for the treatment of HD may provide a benefit over nonselective HTT-lowering approaches, as it aims to preserve the beneficial activities of wtHTT. Targeting a heterozygous single-nucleotide polymorphism (SNP) where the targeted variant is on the mHTT gene is one strategy for achieving allele-selective activity. Herein, we investigated whether stereopure phosphorothioate (PS)- and phosphoryl guanidine (PN)-containing oligonucleotides can direct allele-selective mHTT lowering by targeting rs362273 (SNP3). We demonstrate that our SNP3-targeting molecules are potent, durable, and selective for mHTT in vitro and in vivo in mouse models. Through comparisons with a surrogate for the nonselective investigational compound tominersen, we also demonstrate that allele-selective molecules display equivalent potency toward mHTT with improved durability while sparing wtHTT. Our preclinical findings support the advancement of WVE-003, an investigational allele-selective compound currently in clinical testing (NCT05032196) for the treatment of patients with HD.