Merck
  • Development of a 3D Brain Model to Study Sex-Specific Neuroinflammation After Hemorrhagic Stroke.
Development of a 3D Brain Model to Study Sex-Specific Neuroinflammation After Hemorrhagic Stroke.
Translational stroke research (2024-04-01)
Rezwanul Islam, Hadi Hasan Choudhary, Hritik Mehta, Feng Zhang, Tudor G Jovin, Khalid A Hanafy
ZUSAMMENFASSUNG
Subarachnoid hemorrhage (SAH) accounts for 5% of stroke, with women having a decreased inflammatory response compared to men; however, this mechanism has yet to be identified. One hurdle in SAH research is the lack of human brain models. Studies in murine models are helpful, but human models should be used in conjunction for improved translatability. These observations lead us to develop a 3D system to study the sex-specific microglial and neuroglial function in a novel in vitro human SAH model and compare it to our validated in vivo SAH model. Our lab has developed a 3D, membrane-based in vitro cell culture system with human astrocytes, microglia, and neurons from both sexes. The 3D cultures were incubated with male and female cerebrospinal fluid from SAH patients in the Neuro-ICU. Furthermore, microglial morphology, erythrophagocytosis, microglial inflammatory cytokine production, and neuronal apoptosis were studied and compared with our murine SAH models. The human 3D system demonstrated intercellular interactions and proportions of the three cell types similar to the adult human brain. In vitro and in vivo models of SAH showed concordance in male microglia being more inflammatory than females via morphology and flow cytometry. On the contrary, both in vitro and in vivo models revealed that female microglia were more phagocytic and less prone to damaging neurons than males. One possible explanation for the increased phagocytic ability of female microglia was the increased expression of CD206 and MerTK. Our in vitro, human, 3D cell culture SAH model showed similar results to our in vivo murine SAH model with respect to microglial morphology, inflammation, and phagocytosis when comparing the sexes. A human 3D brain model of SAH may be a useful adjunct to murine models to improve translation to SAH patients.