- Enantiospecific synthesis of genetically encodable fluorescent unnatural amino acid L-3-(6-acetylnaphthalen-2-ylamino)-2-aminopropanoic acid.
Enantiospecific synthesis of genetically encodable fluorescent unnatural amino acid L-3-(6-acetylnaphthalen-2-ylamino)-2-aminopropanoic acid.
The Journal of organic chemistry (2011-07-08)
Zheng Xiang, Lei Wang
PMID21732687
ZUSAMMENFASSUNG
Fluorescent unnatural amino acids (UAAs), when genetically incorporated into proteins, can provide unique advantages for imaging biological processes in vivo. Synthesis of optically pure L-enantiomer of fluorescent UAAs is crucial for their effective application in live cells. An efficient six-step synthesis of L-3-(6-acetylnaphthalen-2-ylamino)-2-aminopropanoic acid (L-Anap), a genetically encodable and polarity-sensitive fluorescent UAA, has been developed. The synthesis takes advantage of a high-yield and enantiospecific Fukuyama-Mitsunobu reaction as the key transformation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
L-ANAP trifluoroacetate, ≥98% (HPLC), powder, fluorescent amino acid analog