Merck
  • Discovery of Non-antibacterial Enrofloxacin Derivatives with Emerging Antiaging Effects through Drug Repurposing and Secondary Development.
Discovery of Non-antibacterial Enrofloxacin Derivatives with Emerging Antiaging Effects through Drug Repurposing and Secondary Development.
Journal of medicinal chemistry (2025-05-09)
Sicong Qiao, Xin Chen, Youyou Chen, Yihe Song, Donglei Shi, Zhifan Mao, Jian Li, Yunyuan Huang, Wenwen Liu
ZUSAMMENFASSUNG
Aging induces dysfunction and increases the risk of chronic diseases in the elderly, positioning the development of antiaging drugs to the forefront of research. Drug repurposing offers an efficient strategy for identifying antiaging lead compounds. In this study, we employed phenotypic screening and discovered that enrofloxacin could extend the lifespan in Caenorhabditis elegans. Based on these findings, we conducted rational drug design to eliminate its antibacterial activity while maintaining the lifespan-extending effect, with the goal of developing safe and novel antiaging compounds. Consequently, JX10 exhibited minimal antibacterial activity and competent antiaging effects in C. elegans, senescent cells, and aged mice. In terms of its mechanism, JX10 acted as a senomorphic agent by suppressing the expression of p38 MAPK and NF-κB. Furthermore, JX10 demonstrated favorable safety and pharmacokinetic properties, supporting the feasibility of JX10 as a promising candidate with the potential for therapeutic interventions in aging and aging-related diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Enrofloxacin hydrochloride, ≥98% (HPLC), powder, Fluoroquinolone antibiotic