Merck
Cytotoxic Effect of Phenylethanoid Glycosides Isolated from Plantago lanceolata L.
Life (Basel, Switzerland) (2023-02-26)
Anna Budzianowska, Ewa Totoń, Aleksandra Romaniuk-Drapała, Małgorzata Kikowska, Jaromir Budzianowski
ZUSAMMENFASSUNG
The aim of the study is to investigate whether the bioactive compounds isolated from P. lanceolata inflorescences, namely, phenylethanoid glucosides, acteoside, plantamajoside, and a flavonoid, isorhamnetin-3-O-rutinoside-4'-O-glucoside, possessed cytotoxic activity against the selected cancer cell lines. The potential antitumor effects of two phenylethanoid glycosides and one flavonoid were evaluated via MTT (3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide) assay on seven human carcinoma cell lines (MCF-7, MDA-MB-231, Caco-2, HepG2, OVCAR-3, U138-MG, U251-MG) and one nontumorigenic mammary epithelial cell line (MCF-12A). For the first time, acteoside was studied in ovarian cancer cell line OVCAR-3, and plantamajoside in all cell lines except breast adenocarcinoma MDA-MB-281 and hepatocarcinoma HepG2. The phenylethanoid glycosides showed stronger cytotoxic activity than that of the glycoside flavonoid. Acteoside and plantamajoside, at concentrations of 200 and 300 μM, respectively, had a highly toxic effect on the selected two cancer cell lines of breast adenocarcinoma MDA-MB-231 and MCF-7, ovarian cancer cell line OVCAR-3, glioblastoma cell line U138-MG, and hepatocarcinoma cell line HepG2. Both glycosides were significantly less cytotoxic towards nontumorigenic cell line MCF-12A; the effect appeared at a concentration of 400 μM. For the first time, the activity of acteoside and plantamajoside was compared in one parallel investigation. The results are discussed against a broad background of existing knowledge on biological effects, their mechanisms, and structure-activity relationships. Phenylethanoids may be potential compounds with cytotoxic activity against the selected cancer types.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Plantamajoside, ≥98% (HPLC)